EMAIL THIS PAGE TO A FRIEND
Environmental toxicology and chemistry
Sensitivity of brain cholinesterase activity to diazinon (BASUDIN 50EC) and fenobucarb (BASSA 50EC) insecticides in the air-breathing fish Channa striata (Bloch, 1793).
PMID 16704077
Abstract
With the expansion of agricultural areas within the Mekong River Delta in Vietnam, a concurrent, dramatic increase has occurred in agrochemical usage. To date, little consideration has been given to the negative impacts of this agricultural activity on the aquatic resources of the region. Both acute toxicity and subacute effects on brain cholinesterase (ChE) of two of the most commonly used insecticides, diazinon and fenobucarb, on adult native snakehead (Channa striata) were evaluated in a static, nonrenewable system, the environmental parameters of which, such as dissolved oxygen, water temperature, and pH, fluctuated similarly to field conditions. Four levels of insecticides, from 0.008 to 0.52 mg/L (for diazinon) and from 0.11 to 9.35 mg/L (for fenobucarb), were tested to assess the effects on the brain ChE activity of the snakehead up to 30 and 10 d for diazinon and fenobucarb, respectively. Diazinon was highly toxic to this fish species, with a 96-h median lethal concentration (LC50) of only 0.79 mg/L, and it also caused long-term ChE inhibition, with activity still significantly inhibited by 30% after 30 d for the three highest concentrations. Fenobucarb was less toxic to this species, with a 96-h LC50 of 11.4 mg/L. Fenobucarb caused more rapid ChE inhibition but also rapid recovery. The results of the present study indicate an urgent need to regulate the usage of these pesticides in the Mekong River Delta.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45488
Fenobucarb, PESTANAL®, analytical standard
C12H17NO2