EMAIL THIS PAGE TO A FRIEND
Organic letters
In pursuit of pestalotiopsin a via zirconocene-mediated ring contraction.
PMID 16706543
Abstract
[reaction: see text] An asymmetric route from the epimeric beta-hydroxy esters 4 and 5 to the densely functionalized (+)-10 and (-)-10, respectively, is described. Either cyclobutanol can be made available as the predominant product. The levorotatory antipode has been transformed into the advanced intermediate 21 bearing side chains destined to become incorporated into the cyclononene ring of the title compound (1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
196215
Bis(cyclopentadienyl)zirconium(IV) dichloride, ≥98%
C10H10Cl2Zr