EMAIL THIS PAGE TO A FRIEND
Biomedical chromatography : BMC
Screening and chromatographic assessing of a novel IgG biomimetic ligand.
PMID 16708378
Abstract
A novel biomimetic ligand, N-benzyloxycarbonyl-l-tyrosine (N-cbz-l-Tyr), was screened by a combination method of molecular docking and immobilized receptor technique. Then, N-cbz-l-Tyr was immobilized on Sepharose CL-4B to prepare a specific affinity adsorbent for immunoglobulin G (IgG). Scatchard analysis of the binding isotherm for IgG on the adsorbent gave an association constant (K(a)) of 4.91 x 10(6) m(-1) and a theoretical maximum adsorption capacity of 17.3 mg IgG/mL gel. IgG with a purity of 98% was separated from human plasma by this new affinity adsorbent.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
CL4B200 Sepharose® CL-4B, Cross-linked