EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
N,N'-carbonyldiimidazole-mediated cyclization of amino alcohols to substituted azetidines and other N-heterocycles.
PMID 16709054
Abstract
Amino alcohols are important synthons for N-heterocycles. We have developed an efficient method to activate hydroxyl groups, which avoids the use of toxic reagents and tolerates a wide variety of functional groups. Our strategy has been applied to the synthesis of functionalized p-methoxyphenyl-protected azetidines, pyrrolidines, and piperidines. The required amino alcohols were synthesized according to an optimized proline-catalyzed Mannich protocol. An azetidine analogue of ezetimibe was synthesized to demonstrate the potential for the synthesis of drug-like molecules.