EMAIL THIS PAGE TO A FRIEND
Clinical and experimental dermatology
Severe cholinergic urticaria successfully treated with scopolamine butylbromide in addition to antihistamines.
PMID 16716168
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O9387 Oxatomide, ≥99%
C27H30N4O