EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
FTIR features of lithium-iron phosphates as electrode materials for rechargeable lithium batteries.
PMID 16716657
Abstract
The essential structural features of lithium-metal phosphates (LMP) have been studied using FTIR spectroscopy which is a sensitive tool to probe the local environment in the solid materials. Various LMP materials where M is iron have been investigated including phospho-olivine LiFePO(4), diphosphate LiFeP(2)O(7), Nasicon-type phosphate Li(3)Fe(2)(PO(4))(3) and dihydrate FePO(4).2H(2)O. Vitreous and amorphous materials are also considered. Analysis of internal and external modes of vibration allows to distinguish between the different phases and the type of cationic environment in the framework. Results corroborate the contribution of the main factors which are responsible for the complexity of the spectra, i.e. departure from ideal symmetry, interactions between polyhedra, bridging atoms and lattice distortion.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
436011
Iron(III) phosphate dihydrate, Fe 29 %
FeO4P · 2H2O