EMAIL THIS PAGE TO A FRIEND
Bioscience, biotechnology, and biochemistry
Identification of 2-(cysteinyl)amido-2-deoxy-D-galacturonic acid residue from the sheath of Leptothrix cholodnii.
PMID 16717434
Abstract
The sheath of Leptothrix cholodnii is a glycoconjugate composed of a polysaccharide and a peptide rich in cysteine. In this study, structural determination of the hydrazinolyzate of the sheath was carried out. Since the hydrazinolyzate is a polysaccharide incorporated with cysteine, it was S-derivatized with a thiol-specific fluorogenic reagent, 4-(aminosulfonyl)-7-fluoro-2,1,3-benzoxadiazole (ABD-F). Fluorescent fragments were purified by HPLC, and their structures were analyzed by mass spectrometry and NMR spectroscopy. The sheath was found to contain 2-(cysteinyl)amido-2-deoxy-D-galacturonic acid residue.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F3639
4-Fluoro-7-sulfamoylbenzofurazan
C6H4FN3O3S