EMAIL THIS PAGE TO A FRIEND
Phytotherapy research : PTR
Cytotoxicity in vitro of naphthazarin derivatives from Onosma arenaria.
PMID 16718737
Abstract
The cytotoxicity of naphthazarin derivatives isolated from the roots of Onosma arenaria on human cervix adenocarcinoma cells (HeLa) and leukaemia K562 cells, as well on non-malignant peripheral blood mononuclear cells (PBMC) was studied. The results show that beta-hydroxyisovalerylalkannin, acetylalkannin and the pigment fraction exhibited high cytotoxicity in vitro against the tested cell lines, as well the healthy PBMC before or after activation with phytohaemagglutinin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
388513
5,8-Dihydroxy-1,4-naphthoquinone, technical grade
C10H6O4