EMAIL THIS PAGE TO A FRIEND
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials
[Analysis of constituents of essential oil from the skin of water caltrop].
PMID 16722314
Abstract
To analyze the constituents of essential oil from the skin of water caltrop. Water steam distillation and GC-MS were used. 58 componds were separated respectively. 56 componds being identified which were 96. 5% of the totle essential oil. Diethyl phthalate, acetamide, N-acetyl-N, N'-1,2-ethanediylbis-, isopropyl palmitate, hexadecanoic acid, Z-11 and octadecanoic acid are the main component of essential oil from the skin of water caltrop.