EMAIL THIS PAGE TO A FRIEND
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica
[Studies on chemical constituents in herb of Myricaria bracteata].
PMID 16722376
Abstract
To study the chemical constituents of Myricaria bracteata. The chemical constituents were isolated by silica gel column chromatography and the structures were elucidated by spectroscopic methods. Eleven compounds were obtained and identified as rhamnetin, 3,5,4'-trihydroxy-7,3'-dimethoxyflavone, 3,5,4'-trihydroxy-7-methoxyflavone, quercetin-3-O-alpha-L-rhamnopyranoside, kaempferol, quercetin, chrysoerio, gallic acid, gallic acid ethylester, beta-sitosterol, daucosterol. All compounds were obtained from M. bracteata for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
17799
Rhamnetin, analytical standard
C16H12O7