EMAIL THIS PAGE TO A FRIEND
Science (New York, N.Y.)
Selective activation of the B natriuretic peptide receptor by C-type natriuretic peptide (CNP).
PMID 1672777
Abstract
The natriuretic peptides are hormones that can stimulate natriuretic, diuretic, and vasorelaxant activity in vivo, presumably through the activation of two known cell surface receptor guanylyl cyclases (ANPR-A and ANPR-B). Although atrial natriuretic peptide (ANP) and, to a lesser extent, brain natriuretic peptide (BNP) are efficient activators of the ANPR-A guanylyl cyclase, neither hormone can significantly stimulate ANPR-B. A member of this hormone family, C-type natriuretic peptide (CNP), potently and selectively activated the human ANPR-B guanylyl cyclase. CNP does not increase guanosine 3',5'-monophosphate accumulation in cells expressing human ANPR-A. The affinity of CNP for ANPR-B is 50- or 500-fold higher than ANP or BNP, respectively. This ligand-receptor pair may be involved in the regulation of fluid homeostasis by the central nervous system.