EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Determination of metoprolol tartrate in tablets and human urine using flow-injection chemiluminescence method.
PMID 16730940
Abstract
In this paper, a simple, rapid and sensitive flow-injection chemiluminescence method has been developed for the determination of metoprolol tartrate, which acts as a kind of sensitizer in the chemiluminescence emission from the redox of SO(3)(2-) with Ce(IV) in acidic medium. Under the optimized conditions, the proposed method allows the measurement of metoprolol tartrate over the range of 1.5 x 10(-8) to 7.3 x 10(-6)mol/L with a detection limit of 4.7 x 10(-9)mol/L (3sigma), and the relative standard deviation for 7.3 x 10(-7)mol/L metoprolol tartrate (n=11) is 2.20%. The utility of this method was demonstrated by determining metoprolol tartrate in tablets and human urine sample.