EMAIL THIS PAGE TO A FRIEND
Diabetes Care
1,5-anhydroglucitol and postprandial hyperglycemia as measured by continuous glucose monitoring system in moderately controlled patients with diabetes.
PMID 16731998
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
54920
(R)-3-Hydroxybutyric acid, ≥98.0% (T)
C4H8O3
34867
1-Butanol, for HPLC, ≥99.7%
C4H10O
360465
1-Butanol, ACS reagent, ≥99.4%
C4H10O
33065
1-Butanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
C4H10O
537993
1-Butanol, 99.9%
C4H10O
901351
1-Butanol, ACS reagent, from renewable source, ≥99.4%
C4H10O
67520
1-Methyl-L-histidine, ≥98.0% (TLC)
C7H11N3O2
166898
3-Hydroxybutyric acid, 95%
C4H8O3
M9005
3-Methyl-L-histidine
C7H11N3O2
W254606
4-Heptanone, ≥97%, FG
C7H14O
101745
4-Heptanone, 98%
C7H14O
270725
Acetone, for HPLC, ≥99.9%
C3H6O
34850
Acetone, for HPLC, ≥99.8%
C3H6O
179124
Acetone, ACS reagent, ≥99.5%
C3H6O
320110
Acetone, ACS reagent, ≥99.5%
C3H6O
673781
Acetone, ACS reagent, ≥99.5%
C3H6O
534064
Acetone, histological grade, ≥99.5%
C3H6O
32201
Acetone, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.5% (GC)
C3H6O
24201
Acetone, puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)
C3H6O
179973
Acetone, Laboratory Reagent, ≥99.5%
C3H6O
L0625
D-(−)-Lactic acid, ≥90%
C3H6O3
295280
Dimethylamine, anhydrous, ≥99%
C2H7N
388246
Dimethylamine solution, 2.0 M in methanol
C2H7N
391956
Dimethylamine solution, 2.0 M in THF
C2H7N
426458
Dimethylamine solution, 40 wt. % in H2O
C2H7N
D1009
Dodecanedioic acid, 99%
C12H22O4
46565
Estriol, VETRANAL, analytical standard
C18H24O3
E-074
Estriol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material
C18H24O3
67028
Methylglyoxal solution, technical, ~40% in H2O
C3H4O2