EMAIL THIS PAGE TO A FRIEND
Organic letters
Total synthesis of (-)-8-O-methyltetrangomycin (MM 47755).
PMID 16737300
Abstract
A stereoselective total synthesis of the natural antibiotic (-)-8-O-methyltetrangomycin 1 is reported. The essential steps for this convergent synthesis are the transformation of a geraniol epoxide into a chiral octadiyne derivative, which was converted into a triyne. The cobalt-mediated [2+2+2] cycloaddition of the triyne led to a benz[a]anthracene system, which was oxidized with Ag(Py)(2)MnO(4) to a benz[a]anthraquinone. Deprotection with aqueous HF in acetonitrile and photooxidation afforded the desired product (-)-1. [reaction: see text]
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00034
6-Deoxy-8-O-methylrabelomycin, ≥95% (LC/MS-ELSD)
C20H16O5