EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Mechanism of Oxidation in the Plant. Part II: An Investigation of Substances which are capable of behaving as Peroxydases.
PMID 16743297
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G8279 Gum guaiac, crude colorimetric peroxidase substrate