EMAIL THIS PAGE TO A FRIEND
Biochemical Society transactions
The reaction of oxidized and reduced glutathione (with some amino-acids) and L-gamma-glutamyl p-nitroanilide with sheep kidney tissue homogenate. An examination of the reaction products by paper chromatography.
PMID 1674706