EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Studies in the biochemistry of micro-organisms: Synthesis of catenarin (1:4:5:7-tetrahydroxy-2-methylanthraquinone), a metabolic product of species of Helminthosporium.
PMID 16747444
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
423742
3,5-Dimethoxytoluene, 98%
C9H12O2