EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
The biochemistry of aromatic amines. 8. Synthesis and detection of di-(2-amino-1-naphthyl) hydrogen phosphate, a metabolite of 2-naphthylamine in dogs.
PMID 16748865
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
161152
2-Nitro-1-naphthol, 95%
C10H7NO3