EMAIL THIS PAGE TO A FRIEND
Phytochemical analysis : PCA
Simultaneous quantification of eudesmanolides and thymol derivatives from tissues of Inula helenium and I. royleana by reversed-phase high-performance liquid chromatography.
PMID 16749422
Abstract
A simple and rapid isocratic reversed-phase high-performance liquid chromatographic method for the quantification of alantolactone/isoalantolactone and three thymol derivatives in roots and root cultures of Inula helenium and I. royleana has been developed. The method could be applied to screen raw materials in search for highly productive plants and in vitro cultures.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML0415
Alantolactone, ≥98% (HPLC)
C15H20O2