EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Synthetic transformation of ptychantin into forskolin and 1,9-dideoxyforskolin.
PMID 16749796
Abstract
Forskolin (1), a highly oxygenated labdane diterpenoid and an activator of adenylate cyclase, has been synthesized in 12 steps and 12% overall yield from ptychantin A (4), which has been isolated from liverwort Ptychanthus striatus in good yield. The 1alpha-hydroxy group was furnished by stereoselective reduction of the corresponding carbonyl group by sodium in t-BuOH. The 9alpha-hydroxy group was introduced stereoselectively by epoxidation of delta(9.11)-enolether. 1,9-Dideoxyforskolin (2), an inhibitor of glucose transporter, has been synthesized in 8 steps and 37% overall yield. The hydroxy group at C-1 was removed by solid-state thicarbonylimidazolation and subsequent radical cleavage.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D3658
1,9-Dideoxyforskolin from Coleus forskohlii, ≥97%, solid
C22H34O5