EMAIL THIS PAGE TO A FRIEND
Journal of Asian natural products research
Chemical constituents from Mahkota dewa.
PMID 16753792
Abstract
A new phenolic glycoside (1), mahkoside A, together with six known compounds including mangiferin (2), kaempferol-3-O-beta-D-glucoside (3), dodecanoic acid (4), palmitic acid (5) ethyl stearate (6) and sucrose (7), were isolated from the pit of Mahkota dewa. Their structures were identified on the basis of spectroscopic analysis. All the compounds were isolated from the title plant for the first time.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W349003
Ethyl stearate, ≥97%
C20H40O2
S8269
Ethyl stearate, ≥99% (capillary GC)
C20H40O2