EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
New triterpenoids isolated from the root bark of Ulmus pumila L.
PMID 16755042
Abstract
Three new triterpenoids, 1-3, were isolated from the dried root bark of Ulmus pumila. Along with the three new compounds, six known triterpenoids, epifriedelanol (4), friedelin (5), oleanolic acid (6), maslinic acid (7), camaldulenic acid (8), and arjunolic acid (9) were also isolated. The structures of new compounds were established as ulmudiol (bauer-7-ene-1alpha,3beta-diol, 1), dehydroulmudiol [bauer-7,9(11)-diene-1alpha,3beta-diol, 2], and ulmuestone [3alpha-hydroxy-11alpha-(4'-hydroxy-3'-methoxy)benzoyloxybauer-1-one, 3], on the basis of extensive 1D and 2D NMR spectroscopic data interpretation. In addition, the cytotoxic activities of these compounds are also reported.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SMB00119
Arjunolic acid, ≥95% (LC/MS-ELSD)
C30H48O5