EMAIL THIS PAGE TO A FRIEND
Journal of cosmetic science
Malassezia and seborrheic dermatitis: etiology and treatment.
PMID 16758556
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
257710
Selenium sulfide
S2Se