EMAIL THIS PAGE TO A FRIEND
Chemosphere
"Does atrazine transform to propazine in aquatic sediments?": Comment on "biological and chemical transformations of atrazine in coastal aquatic sediments" by Kelly L. Smalling and C. Marjorie Aelion [Chemosphere 62 (2006) 188-196].
PMID 16762390