EMAIL THIS PAGE TO A FRIEND
Advances in experimental medicine and biology
Glutathione conjugation as a mechanism of targeting latent quinones to the kidney.
PMID 1676871
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
167134
Bromohydroquinone, 97%
C6H5BrO2