EMAIL THIS PAGE TO A FRIEND
Chemical reviews
Chemistry of 2-acetyl-1-pyrroline, 6-acetyl-1,2,3,4-tetrahydropyridine, 2-acetyl-2-thiazoline, and 5-acetyl-2,3-dihydro-4H-thiazine: extraordinary Maillard flavor compounds.
PMID 16771451
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W381718
2-Acetyl-2-thiazoline, ≥96%, FG
C5H7NOS