EMAIL THIS PAGE TO A FRIEND
Teratogenesis, carcinogenesis, and mutagenesis
A genotoxicological study of hexachlorobenzene and pentachloroanisole.
PMID 1677498
Abstract
The potential mutagenic activity of hexachlorobenzene (HCB) and pentachloroanisole (PCA) was investigated. No genotoxicity after application on Salmonella typhimurium (Ames test), Escherichia coli, and human peripheral blood lymphocytes in vitro was observed.