EMAIL THIS PAGE TO A FRIEND
Biotechnology and applied biochemistry
Optimization of the conjugation method for a serogroup B/C meningococcal vaccine.
PMID 16776648
Abstract
A conjugate meningococcal vaccine against serogroup B/C consisting of capsular PS (polysaccharide) from serogroup C conjugated to OMV (outer membrane vesicle) from serogroup B would be a very useful vaccine in regions where there is a prevalence of both serogroups, for example in Brazil. For this purpose, the conjugation method that uses ADHy (adipic acid dihydrazide) as spacer and a carbodi-imide derivative, EDAC [1-ethyl-3-(3-dimethylaminopropyl)carbodi-imide], as catalyser was optimized looking for synthesis yield and maintenance of the antigenicity of both components. The best synthesis conditions preserving the vaccine immunogenicity resulted in a final yield of approx. 17%. Immunogenicity of the vaccine was highest when 10% of the sialic acid residues of the PS were occupied by the ADHy spacer. Sterilization of the conjugate by filtration through a 0.22-microm-pore-size membrane resulted in a low recovery of protein and PS (approximately 50%), although the vaccine immunogenicity was maintained. Using gamma irradiation on freeze-dried sample, it was possible to maintain the integrity of OMV structure and, consequently, its ability to induce bactericidal antibodies.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0638
Adipic acid dihydrazide, ≥98% (titration)
C6H14N4O2