EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
The use of acetylated ferricytochrome c for the detection of superoxide radicals produced in biological membranes.
PMID 167777
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C4186
Cytochrome c partially acetylated from equine heart, lyophilized powder