EMAIL THIS PAGE TO A FRIEND
Journal of AOAC International
Determination of semicarbazide in baby food by liquid chromatography/tandem mass spectrometry: interlaboratory validation study.
PMID 16792072
Abstract
An interlaboratory validation study funded by the European Commission, Directorate General for Health and Consumer Protection (DG SANCO), was conducted to evaluate the effectiveness of a liquid chromatography/tandem mass spectrometry (LC/MS/MS) method for the determination of semicarbazide (SEM) in different types of baby food at a possible future European regulatory limit (10 ng/g). The test portion of the sample was extracted with hydrochloric acid, and the analyte was derivatized with 2-nitrobenzaldehyde, with 1,2-[15N2, 13C] SEM as an internal standard. The extract was neutralized and then purified on a solid-phase extraction cartridge. The SEM was determined by reversed-phase LC with detection by MS/MS. Apple puree, rice pudding, and meat/vegetable meal baby food materials, spiked with SEM at levels of about 3, 10, and 30 ng/g, respectively, were sent to 20 laboratories in 12 different European countries, which submitted results from 17 participants. Recoveries ranged from 88.8 to 106.1%. Based on results for spiked samples (blind pairs at 3 levels), the relative standard deviations for repeatability (RSDr) ranged from 4.2 to 6.9% and the relative standard deviations for reproducibility (RSDR) ranged from 16.6 to 24.3%. The method showed acceptable within- and between-laboratory precision for all 3 matrixes, as evidenced by HorRat values, at the target levels for the determination of SEM.
Related Materials