EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Single-step conversion of dimethyl terephthalate into cyclohexanedimethanol with Ru5PtSn, a trimetallic nanoparticle catalyst.
PMID 16795100
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
125598
1,4-Cyclohexanedimethanol, mixture of cis and trans, 99%
C8H16O2