EMAIL THIS PAGE TO A FRIEND
Biopolymers
Solvent-induced lysozyme gels: effects of system composition and temperature on structural and dynamic characteristics.
PMID 16802304
Abstract
The gelation process of lysozyme in water/tetramethylurea in the presence of salt was investigated as a function of temperature and system composition by rheology, infrared spectroscopy, and microcalorimetry. Times and temperatures of gelation were determined from the variation of the storage (G') and loss (G'') moduli. It was found that gelation times follow exponential decays with both protein and tetramethylurea (TMU) concentrations and with temperature. The activation energy for the overall process shows a linear dependence on TMU mass fraction. A strongly increased beta-sheet content and reduced alpha-helix occur with the increase of TMU concentration in the binary solvent. Also, a linear decrease of lysozyme denaturation temperature and enthalpy on TMU concentration is found for the TMU mass fraction up to 0.5, above which no denaturation signal can be detected.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T24503
Tetramethylurea, 99%
C5H12N2O