EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
2-azaadamantane N-oxyl (AZADO) and 1-Me-AZADO: highly efficient organocatalysts for oxidation of alcohols.
PMID 16802802
Abstract
Development of a stable nitroxyl radical class of catalysts, 2-azaadamantane N-oxyl (AZADO) and 1-Me-AZADO, for highly efficient oxidation of alcohols is described. AZADO and 1-Me-AZADO exhibit superior catalytic proficiency to TEMPO, converting various sterically hindered alcohols to the corresponding carbonyl compounds in excellent yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
701718
2-Azaadamantane-N-oxyl, 90%
C9H14NO