EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Sorbent extraction of rubeanic acid-metal chelates on a new adsorbent: Sepabeads SP70.
PMID 16806687
Abstract
A sorbent extraction procedure for lead, iron, cadmium and manganese ions on Sepabeads SP70 adsorption resin has been presented prior to their flame atomic absorption spectrometric determinations. By the passage of aqueous samples including analyte ions-rubeanic acid chelates through Sepabeads SP70 column, metal chelates adsorb quantitatively and almost all matrix elements will pass through the column to drain. The influence of potential interfering ions was also studied. The validation of the method was made though the analysis of LGC 6010 Hard drinking water, SRM 1577b Bovine liver and GBW 07603 Bush branches and leaves standard reference materials (SRM). The method was applied to the determination of analyte ions from various water, wastewater, cow meat and milk, red wine, and tobacco samples with successfully results.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
379387
Dithiooxamide, 97%
C2H4N2S2
13962-U Sepabeads® SP70, bucket of 1000 g