EMAIL THIS PAGE TO A FRIEND
Reproductive toxicology (Elmsford, N.Y.)
Prenatal developmental toxicity study of the basic rubber accelerator, 1,3-di-o-tolylguanidine, in rats.
PMID 16806806
Abstract
Pregnant rats were given 1,3-di-o-tolylguanidine (DTG) by gavage at 0, 10, 20 or 40 mg/kg bw/day on days 6-19 of pregnancy and the pregnancy outcome was determined on day 20 of pregnancy. At 40 mg/kg bw/day, deaths were observed in four out of 24 females. The incidences of females showing mydriasis at 20 and 40 mg/kg bw/day and showing decreased locomotor activity at 40 mg/kg bw/day were significantly increased. Alopecia, bradypnea, prone position and tremor were also observed at mg/kg bw/day. The maternal body weight gain at 20 and 40 mg/kg bw/day and food consumption at 40 mg/kg bw/day were significantly reduced. A significantly decreased weight of the gravid uterus, increased incidence of postimplantation loss, decreased number of live fetuses, and lowered weights of fetuses and placentae were found at 40 mg/kg bw/day. The incidences of the total number of fetuses with external malformations at 40 mg/kg bw/day and with skeletal malformations at 20 and 40 mg/kg bw/day were significantly increased. Significantly higher incidences of fetuses with brachydactyly and short tail and defects of caudal vertebrae, phalanges and metacarpals were observed at 40 mg/kg bw/day. Delayed ossification was also noted at 40 mg/kg bw/day. The data indicate that DTG is teratogenic at maternal toxic doses and the NOAELs of DTG for maternal and developmental toxicity are 10 mg/kg bw/day in rats.