EMAIL THIS PAGE TO A FRIEND
Molecular biology reports
Cloning and characterization of a novel KRAB-domain-containing zinc finger gene (ZNF284L).
PMID 16817023
Abstract
The zinc finger gene (ZNF) family plays an important role in the regulation of transcription. This study reports the cloning and characterization of a novel human zinc finger protein cDNA (ZNF284L) from fetal brain cDNA library. The ZNF284L cDNA is 2223 bp in length encoding a 593-aa polypeptide. The protein contains a KRAB A+b box and eleven C2H2 type zinc finger motifs. ZNF284L gene is mapped to 19q13.2-19q13.3 with 5 exons, and the expression pattern of ZNF284L gene was also examined by reverse transcription polymerase chain reaction (RT-PCR). The transcripts were detected in the human lung, liver, pancreas, thymus, heart, placenta, spleen, prostate, ovary, small intestine and colon, but in human brain, skeletal muscle, kidney, testis and peripheral blood leukocyte, no expression was detected.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV50430
Anti-ZNF284 antibody produced in rabbit, affinity isolated antibody