EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Toward the total synthesis of spirastrellolide A. Part 1: Strategic considerations and preparation of the southern domain.
PMID 16823782