EMAIL THIS PAGE TO A FRIEND
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy
Selective thiourea optical probe based on thiourea-induced removal of chloroacetyl group from chloroacetylamine.
PMID 16824787
Abstract
A novel chromogenic probe for thiourea, N-chloroacetyl parafuchsin (CAP), was designed and synthesized. The method was based on the reaction of CAP with thiourea, resulting in the initial formation of the S-substituted thioformamidine hydrochloride, which would then undergo the intramolecular amidinolysis to liberate the corresponding amine, and thus leading to an absorbance increase at visible spectral range. Based on this mechanism, a highly selective optical probe for thiourea was developed. Under optimal conditions, absorbance increase (DeltaA) at 562 nm is proportional to thiourea concentration up to 0.2 mmolL(-1) with a detection limit of 2.1 micromolL(-1) (3sigma). Because of the selective removal of chloroacetyl group from CAP by thiourea, there is little interference by other molecules.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0267
2-Chloroacetamide, ≥98%
C2H4ClNO
22790
2-Chloroacetamide, ≥98.0% (HPLC)
C2H4ClNO