EMAIL THIS PAGE TO A FRIEND
Acta odontologica Scandinavica
Optimizing resin cement removal around esthetic crown margins.
PMID 16829499
Abstract
To quantify luting cement at the crown-tooth interface of esthetic crowns fabricated using four different techniques and two methods of excess cement removal. Four methods of crown fabrication were used: the feldspathic porcelain and platinum foil technique, the feldspathic porcelain and refractory die technique, the resin composite crown and CAD/CAM technique, and the feldspathic porcelain crown and CAD/CAM technique. Half of the cemented crowns were allocated to Group A: removal of excess cement by flicking-off with a plastic instrument 3 min after initial polymerization, or Group B: removal of excess cement using a wiping action with cotton pellets. Morphologic measurements, using non-destructive digital profilometry, were made of the volume of excess cement (mm(3)), mean maximum and mean depth of excess cement (microm), and surface area of excess cement (mm(2)). ANOVA and Duncan post-hoc tests revealed no statistical differences (p<0.05) between the four types of crown fabrication with respect to volume and mean depth of retained luting cement. There was a significantly greater volume, mean depth, and mean maximum depth of luting cement retained using the "flick off" method compared to the cotton pellet "wiping" method for excess cement removal, but no statistical differences in mean surface area between the two methods. Following removal of excess luting cement, as judged clinically, using two methods, subclinical amounts of cement remained adherent to the tooth surface of all specimens at the crown-tooth interface.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
GF41858309
Platinum, foil, 0.2m coil, thickness 0.05mm, coil width 1mm, as rolled, 99.99%
Pt
GF58445433
Platinum, foil, 0.2m coil, thickness 0.125mm, coil width 2.5mm, as rolled, 99.99%
Pt
GF34340408
Platinum, foil, 0.2m coil, thickness 0.15mm, coil width .9mm, as rolled, 99.99%
Pt
GF09598809
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width .4mm, as rolled, 99.95%
Pt
GF07973961
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width .85mm, as rolled, 99.95%
Pt
GF65809222
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 1.2mm, as rolled, 99.95%
Pt
GF35123307
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 1.6mm, as rolled, 99.95%
Pt
GF21169526
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 2mm, as rolled, 99.95%
Pt
GF13420257
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 5mm, as rolled, 99.95%
Pt
GF60594442
Platinum, foil, 0.5m coil, thickness 0.025mm, coil width 8mm, as rolled, 99.95%
Pt
GF52150801
Platinum, foil, 0.5m coil, thickness 0.029mm, coil width .34mm, annealed, 99.99%
Pt
GF91778024
Platinum, foil, 0.5m coil, thickness 0.030mm, coil width .7mm, as rolled, 99.95%
Pt
GF25102897
Platinum, foil, 0.5m coil, thickness 0.045mm, coil width .75mm, as rolled, 99.95%
Pt
GF04267795
Platinum, foil, 0.5m coil, thickness 0.05mm, coil width 1mm, as rolled, 99.99%
Pt
GF66494795
Platinum, foil, 1.2mm disks, thickness 0.025mm, as rolled, 99.95%
Pt
GF86033167
Platinum, foil, 10mm disks, thickness 0.004mm, as rolled, 99.95%
Pt
GF06751267
Platinum, foil, 10mm disks, thickness 0.005mm, as rolled, 99.95%
Pt
GF96551354
Platinum, foil, 10mm disks, thickness 0.0075mm, as rolled, 99.95%
Pt
GF73706058
Platinum, foil, 10mm disks, thickness 0.0125mm, as rolled, 99.95%
Pt
GF29522101
Platinum, foil, 10mm disks, thickness 0.01mm, as rolled, 99.95%
Pt
GF31105864
Platinum, foil, 10mm disks, thickness 0.025mm, as rolled, 99.95%
Pt
GF06396532
Platinum, foil, 10mm disks, thickness 0.025mm, as rolled, 99.99+%
Pt
GF61547632
Platinum, foil, 8mm disks, thickness 0.01mm, as rolled, 99.95%
Pt
GF11448840
Platinum, foil, 8mm disks, thickness 0.025mm, as rolled, 99.95%
Pt
GF60675442
Platinum, foil, 8mm disks, thickness 0.05mm, as rolled, 99.95%
Pt
GF38543946
Platinum, foil, 8mm disks, thickness 0.125mm, as rolled, 99.95%
Pt
GF65307090
Platinum, foil, 8mm disks, thickness 0.125mm, as rolled, 99.99+%
Pt
GF05324183
Platinum, foil, 8mm disks, thickness 0.1mm, as rolled, 99.99+%
Pt
GF40155533
Platinum, foil, light tested, 25x25mm, thickness 0.005mm, as rolled, 99.95%
Pt
GF02102948
Platinum, foil, light tested, 25x25mm, thickness 0.025mm, as rolled, 99.99+%
Pt