EMAIL THIS PAGE TO A FRIEND
Se pu = Chinese journal of chromatography
[Chiral separation of dipeptides by capillary electrophoresis].
PMID 16830473
Abstract
Peptides are increasingly used as pharmaceutical agents. Many small peptides are the essential compounds in biological systems. Direct chiral separation of dipeptide derivatives using 9-(2-carbazole) ethyl chloroformate (CEOC) as the derivatizing agent by capillary electrophoresis (CE) with beta-cyclodextrin (beta-CD) and sodium deoxycholate (SDC) as chiral selectors has been developed. It has been well recognized that the combination of the binary selectors can enhance the selectivity and resolution instead of either beta-CD or SDC alone. The molar ratio of the binary chiral selectors, the buffer concentration and pH of Tris-H3PO4, organic modifier were studied and optimized. Complete chiral separations for 14 dipeptide derivatives using beta-CD and SDC as binary chiral selectors were obtained. The results indicated that each pair of D/L chiral resolution was more than 3.63, and the highest resolution for Gly-Ala was 43.14.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0878
Ala-Gly
C5H10N2O3
50150
Gly-Ala, ≥99.0% (NT)
C5H10N2O3