EMAIL THIS PAGE TO A FRIEND
Journal of the American Academy of Child and Adolescent Psychiatry
Homatropine and psychosis in Weill-Marchesani syndrome.
PMID 16832313
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H0800000
Homatropine hydrobromide, European Pharmacopoeia (EP) Reference Standard
C16H21NO3 · HBr