EMAIL THIS PAGE TO A FRIEND
Bulletin of environmental contamination and toxicology
Persistence and dissipation of O-sec-butylphenyl N-methylcarbamate (BPMC) in rice ecosystem.
PMID 16832766
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45488
Fenobucarb, PESTANAL®, analytical standard
C12H17NO2