EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. A
Time-resolved chemically induced dynamic nuclear polarization studies of structure and reactivity of methionine radical cations in aqueous solution as a function of pH.
PMID 16833344
Abstract
Using time-resolved chemically induced dynamic nuclear polarization (CIDNP) techniques, we have studied the mechanism of the photoreactions of triplet excited 4-carboxybenzophenone (CBP) with l-methionine (Met) and 3-(methylthio)propylamine (MTPA) in aqueous solution and the details of the formation of CIDNP at pH from 6.7 to 13.6. At a pH below the pKa of the nitrogen atom of Met, the CIDNP is strongly affected by degenerate electron exchange between the S-S cationic radical dimer and the zwitterionic form of Met with the rate constant kex = 3.4 x 10(8) s(-1) providing an exhaustive explanation of the pH dependence of steady-state CIDNP that was previously interpreted as a manifestation of fast interconversion among three different methionine radical species (Goez, M.; Rozwadowski, J. J. Phys. Chem. A 1998, 102, 7945-7953). By analyzing the polarization of different protons formed in geminate recombination as a function of the pH, we obtained the branching ratio between two reaction pathways for oxidative quenching of (T)CBP via electron transfer from the sulfur and nitrogen atoms of Met and MTPA. Nuclear spin-lattice relaxation times were determined in the dimeric cation radical of Met (T1,S = 8.5 micros). In the cyclic radical cation of MTPA with a three-electron two-center S-N bond, the estimated paramagnetic relaxation is comparatively slow for all protons. Fast deprotonation of the primary aminium radical cation of MTPA and Met in strongly basic solution takes place on the submicrosecond time scale leading to efficient formation of CIDNP in the neutral aminyl radical.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
639095
3-(Methylthio)propylamine, 97%
C4H11NS