EMAIL THIS PAGE TO A FRIEND
American Journal of Physiology: Renal Physiology
Mineralocorticoid receptor blockade confers renoprotection in preexisting chronic cyclosporine nephrotoxicity.
PMID 16835406