EMAIL THIS PAGE TO A FRIEND
Nucleosides, nucleotides & nucleic acids
Synthesis of 6-arylthio analogs of 2',3'-dideoxy-3'-fluoroguanosine and their effect against hepatitis B virus replication.
PMID 16838853
Abstract
A key compound, 2-amino-6-chlor-9-(2,3-dideoxy-3-fluoro-beta-D-erythro-pentofuranosyl)puine, was prepared from 2-amino-6-chloropurine riboside in 5 steps, then subjected to the nucleophilic displacement with benzenethiols to afford 6-arylthio congeners. These compounds showed a similar anti-HBV effect to that of 2',3' dideoxy-3'-fluoroguanosine.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4634
2-Amino-6-chloropurine riboside, ≥95%
C10H12ClN5O4