EMAIL THIS PAGE TO A FRIEND
Bulletin of experimental biology and medicine
Pharmacokinetics of amixin after repeated peroral administration to mice.
PMID 16848231
Abstract
Pharmacokinetics of amixin was studied after repeated administration (5 days) to animals. Perorally administered amixin is characterized by high bioavailability and is present in the circulation in high concentrations for a long time. The main pharmacokinetic parameters were estimated by the method of linear regression because of slow elimination of amixin from organs and tissues. Our results indicate that repeated treatment with amixin holds much promise for the prevention and therapy of chronic diseases (particularly hepatitides).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
220957
Tilorone dihydrochloride, 95%
C25H34N2O3 · 2HCl