EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Ambient pressure syntheses of size-controlled corundum-type In2O3 nanocubes.
PMID 16848456
Abstract
Hydrolysis of In(O-iPr)3 by 10 molar excess of water at 90 degrees C in a surfactant/solvent mixture of oleylamine/oleic acid/trioctylamine provides very small nanoparticles (<5 nm in diameter) of In(O)(OH). Subsequent in situ thermolysis of the formed In(O)(OH) nanoparticles at 350 degrees C and ambient pressure produces monodisperse h-In2O3 nanocubes, which can form an extended two-dimensional array on a flat surface. The size of the In2O3 nanocubes (8, 10, and 12 nm) could be easily controlled by the simple change in the amounts of employed surfactants. The h-In2O3 nanocube samples show blue PL emissions at room temperature due to, presumably, systematic oxygen vacancy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
760285
Indium(III) isopropoxide solution, 5 % (w/v) in isopropanol
C9H21InO3