EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Simulation of scanning tunneling microscope images of 1,3-cyclohexadiene bound to a silicon surface.
PMID 16851118
Abstract
Scanning tunneling microscope (STM) images of 1,3-cyclohexadiene bound to silicon are interpreted using a nonequilibrium Green's function method. The resolution of the carbon-carbon double bond for positive bias voltages but not for negative bias voltages is explained using a quasiprobability density analysis. The asymmetry in the images arises from the system's voltage dependent electronic structure. A pi* orbital is found to be responsible for the empty state STM images of the carbon-carbon double bond, which is observed experimentally. The pi orbital relevant for the opposite bias does not produce an STM image sharply localized in the bond region because the molecule induces a Si-surface dipole dependent on the bias. The dipole voltage dependence arises from molecular charging. This result emphasizes the importance of simulating the molecule as an element in an open quantum system.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C100005
1,3-Cyclohexadiene, contains 0.05% BHT as inhibitor, 97%
C6H8