EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Studies on electrochemical oxidation mechanisms of polyamides containing N-methylpyrrole and N-methylimidazole by electrospray ionization tandem mass spectrometry.
PMID 16853189
Abstract
Electrochemical oxidization mechanism of polyamides containing N-methylpyrrole and N-methylimidazole was investigated by electrospray ionization tandem mass spectrometry (ESI-MS(n)). The spectra of the oxidization products unambiguously revealed that one or two oxygen atoms are added to the polyamide after the bulk electrolysis. It was found that electrochemical oxidation preferably takes place on the imidazole ring of the polyamide with both imidazole and pyrrole to produce a carbonyl group on the rings. ESI tandem mass spectrometry has proven to be an excellent method for the structural identification of electrochemical oxidation products of DNA-recognizing polyamides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W509973
1-Methylpyrrole, ≥99%
C5H7N
M78801
N-Methylpyrrole, 99%
C5H7N