EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Synthesis of TiO2 photocatalysts in supercritical CO2 via a non-hydrolytic route.
PMID 16853417
Abstract
Nanoscaled TiO2 powders with narrow size dispersion were prepared in supercritical carbon dioxide via non-hydrolytic acylation/deacylation of titanium alkoxide precursors with or without tris-fluorination. The microstructures of these powders were characterized by spectroscopic (FTIR, TGA, and XRD), microscopic (SEM or TEM), and surface area (BET) measurements. Photocatalytic oxidation of 1-octanol on these calcined TiO2 powders and on commercial T805 TiO2 suspended in aerated supercritical carbon dioxide revealed relative reactivity controlled by the powder microstructures. Calcined TiO2 prepared from titanium(IV) isopropoxide and trifluoroacetic anhydride was effectively dispersed in aerated supercritical carbon dioxide under stirring and exhibited high photocatalytic oxidation activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
106232
Trifluoroacetic anhydride, ReagentPlus®, ≥99%
C4F6O3